C12H316CE3A9
Chemcia Scientific, LLC
1-(4-Methyl-2-nitro-phenyl)-propan-2-one
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
1-(4-methyl-2-nitrophenyl)acetone
Inchi Code
1S/C10H11NO3/c1-7-3-4-9(6-8(2)12)10(5-7)11(13)14/h3-5H,6H2,1-2H3
InChI key
ACLPRWHHZSVIRR-UHFFFAOYSA-N
storage temp.
0-5C
Purity
92%
Country of Origin
US
Shipping Temperature
Ambient
Physical Form
Red Liquid
Product Link
http://www.chemcia.com/product_fl02.asp?id=534
MSDS
http://www.chemcia.com/product_fl02.asp?id=534
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service