Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CH4H5F6D88D8

ChemBridge Corporation

methyl 3,3-difluoro-2-oxopropanoate

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
138.07
MDL number:

Iupac Name

methyl 3,3-difluoro-2-oxopropanoate

Inchi Code

1S/C4H4F2O3/c1-9-4(8)2(7)3(5)6/h3H,1H3

InChI key

NTVXPNNVBNHMOK-UHFFFAOYSA-N

storage temp.

-20C

Purity

95%

Country of Origin

KZ

Shipping Temperature

RT

Physical Form

liquid

Product Link

https://www.hit2lead.com/building-blocks/4041032

MSDS

https://www.hit2lead.com/sds/4041032_sds.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service