Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CH4454669682

ChemBridge Corporation

tert-butyl rac-(3aR,6aS)-4-oxohexahydrocyclopenta[c]pyrrole-2(1H)-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
225.29
MDL number:

Iupac Name

tert-butyl (3aR,6aS)-4-oxohexahydrocyclopenta[c]pyrrole-2(1H)-carboxylate

Inchi Code

1S/C12H19NO3/c1-12(2,3)16-11(15)13-6-8-4-5-10(14)9(8)7-13/h8-9H,4-7H2,1-3H3/t8-,9+/m1/s1

Inchi Key

MZQZNAQWIOWKSR-BDAKNGLRSA-N

Storage temp.

RT

Purity

95%

Country of Origin

KZ

Shipping Temperature

RT

Physical Form

liquid

Product Link

https://www.hit2lead.com/building-blocks/4079226

MSDS

https://www.hit2lead.com/sds/4079226_sds.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service