CH4424845569
ChemBridge Corporation
[2-(dimethylamino)phenyl]boronic acid hydrochloride hydrate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Molecular Weight:
219.48
MDL number:
Iupac Name
2-(dimethylamino)phenylboronic acid hydrochloride hydrate
Inchi Code
1S/C8H12BNO2.ClH.H2O/c1-10(2)8-6-4-3-5-7(8)9(11)12;;/h3-6,11-12H,1-2H3;1H;1H2
InChI key
NCICGZYBMKVZGR-UHFFFAOYSA-N
storage temp.
-20C
Salt Data
HCl H2O
Purity
95%
Country of Origin
KZ
Shipping Temperature
RT
Physical Form
solid
Product Link
https://www.hit2lead.com/building-blocks/4008672
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://www.hit2lead.com/sds/4008672_sds.pdf
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service