Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CH4182075514

ChemBridge Corporation

2-(4H-1,2,4-triazol-4-yl)isonicotinic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
190.16
MDL number:

Iupac Name

2-(4H-1,2,4-triazol-4-yl)isonicotinic acid

Inchi Code

1S/C8H6N4O2/c13-8(14)6-1-2-9-7(3-6)12-4-10-11-5-12/h1-5H,(H,13,14)

InChI key

SXTXKDSUUOPOJA-UHFFFAOYSA-N

storage temp.

RT

Purity

95%

Country of Origin

KZ

Shipping Temperature

RT

Physical Form

solid

Product Link

https://www.hit2lead.com/building-blocks/4017324

MSDS

https://www.hit2lead.com/sds/4017324_sds.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service