Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATC311763398

Atlantic Research Chemicals Ltd

Benzyl 2-Formylpyrrolidine-1-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
233.27
MDL number:
Pricing and availability is not currently available. Contact Technical Service

Iupac Name

benzyl 2-formyl-1-pyrrolidinecarboxylate

Inchi Code

1S/C13H15NO3/c15-9-12-7-4-8-14(12)13(16)17-10-11-5-2-1-3-6-11/h1-3,5-6,9,12H,4,7-8,10H2

InChI key

ASNHSGGMNWXBEI-UHFFFAOYSA-N

Purity

95

Country of Origin

GB

Product Link

https://atlantic-chemicals.com/product/?code=AH04501

MSDS

https://atlantic-chemicals.com/product/?code=AH04501


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service