Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATEH99180594

AstaTech, Inc.

SE-PHENYL-L-SELENOCYSTEINE

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$350.00
100 MG
$873.75

Vendor SKU: C16268-25MG

$350.00


Ships in 10 days - from Aldrich Partner.


Select a Size

Change View
25 MG
$350.00
100 MG
$873.75

About This Item

CAS Number:
Molecular Weight:
244.14
MDL number:

Vendor SKU: C16268-25MG

$350.00


Ships in 10 days - from Aldrich Partner.

Iupac Name

(2R)-2-amino-3-(phenylselanyl)propanoic acid

Inchi Code

1S/C9H11NO2Se/c10-8(9(11)12)6-13-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)/t8-/m0/s1

InChI key

PFLVUZBNZZOUSS-QMMMGPOBSA-N

Purity

95%

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=157200

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=C16268


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service