Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATEH98C472E4

AstaTech, Inc.

METHYL 3-(AZETIDIN-3-YL)PROPANOATE HCL

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$276.25
250 MG
$708.75
1 G
$2,121.25

Vendor SKU: AT26042-50MG

$276.25


Availability
Ships in 10 days - from Aldrich Partner.


Select a Size

Change View
50 MG
$276.25
250 MG
$708.75
1 G
$2,121.25

About This Item

CAS Number:
Molecular Weight:
179.65
MDL number:

Vendor SKU: AT26042-50MG

$276.25


Availability
Ships in 10 days - from Aldrich Partner.

Iupac Name

methyl 3-(azetidin-3-yl)propanoate hydrochloride

Inchi Code

1S/C7H13NO2.ClH/c1-10-7(9)3-2-6-4-8-5-6;/h6,8H,2-5H2,1H3;1H

InChI key

RPXVGLQNMJTESA-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=225119

Signal Word

Warning

Hazard Statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=AT26042


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service