Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATEH97E805ED

AstaTech, Inc.

N-ETHYL-2-METHYLBENZENE-1-SULFONAMIDE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
199.27
MDL number:

Iupac Name

N-ethyl-2-methylbenzenesulfonamide

Inchi Code

1S/C9H13NO2S/c1-3-10-13(11,12)9-7-5-4-6-8(9)2/h4-7,10H,3H2,1-2H3

InChI key

NATWUQFQFMZVMT-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

Pale-yellow to Yellow-brown Liquid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=210787

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=D88149


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service