Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATEH96F12508

AstaTech, Inc.

METHYL 4-BROMO-2,3-DIFLUORO-5-IODOBENZOATE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
376.92
MDL number:

Iupac Name

methyl 4-bromo-2,3-difluoro-5-iodobenzoate

Inchi Code

1S/C8H4BrF2IO2/c1-14-8(13)3-2-4(12)5(9)7(11)6(3)10/h2H,1H3

InChI key

CSBWTYPGEVRENA-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=124378

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=W15359


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service