Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATEH93EDCCCB

AstaTech, Inc.

2-BROMO-1-(5-BROMOFURAN-2-YL)ETHANONE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
267.9
MDL number:

Iupac Name

2-bromo-1-(5-bromo-2-furyl)ethanone

Inchi Code

1S/C6H4Br2O2/c7-3-4(9)5-1-2-6(8)10-5/h1-2H,3H2

InChI key

WTFKIDCAQZPAHP-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

Yellow to Brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=172825

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=C90418


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service