Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATEH75DC01DB

AstaTech, Inc.

9-HYDROXYMETHYL-2-(MALEIMIDOPROPIONYLAMINO)FLUORENE

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$411.25

Vendor SKU: C90951-250MG

$411.25


Ships in 10 days - from Aldrich Partner.


Select a Size

Change View
250 MG
$411.25

About This Item

CAS Number:
Molecular Weight:
362.38
MDL number:

Vendor SKU: C90951-250MG

$411.25


Ships in 10 days - from Aldrich Partner.

Iupac Name

3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)-N-(9-(hydroxymethyl)-9H-fluoren-2-yl)propanamide

Inchi Code

1S/C21H18N2O4/c24-12-18-15-4-2-1-3-14(15)16-6-5-13(11-17(16)18)22-19(25)9-10-23-20(26)7-8-21(23)27/h1-8,11,18,24H,9-10,12H2,(H,22,25)

InChI key

PVNUHVREGVTVKF-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

Pale-yellow to Yellow-brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=173371

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=C90951


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service