Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATEH3031F0B5

AstaTech, Inc.

5-IODO-2-(TRIFLUOROMETHOXY)PYRIDINE

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$250.00
250 MG
$596.25
1 G
$1,721.25

Vendor SKU: 98367-50MG

$250.00


Ships in 10 days - from Aldrich Partner.


Select a Size

Change View
50 MG
$250.00
250 MG
$596.25
1 G
$1,721.25

About This Item

CAS Number:
Molecular Weight:
289
MDL number:

Vendor SKU: 98367-50MG

$250.00


Ships in 10 days - from Aldrich Partner.

Iupac Name

5-iodo-2-(trifluoromethoxy)pyridine

Inchi Code

1S/C6H3F3INO/c7-6(8,9)12-5-2-1-4(10)3-11-5/h1-3H

InChI key

GFWZFVCJFBKZFK-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

Light-red to Brown Liquid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=145920

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=98367


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service