Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATEH0DDC42F4

AstaTech, Inc.

(4-(CHLOROMETHYL)PHENYL)(3,4-DIHYDROISOQUINOLIN-2(1H)-YL)METHANONE

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$180.00
100 MG
$471.25
250 MG
$873.75

Vendor SKU: F20138-25MG

$180.00


Ships in 10 days - from Aldrich Partner.


Select a Size

Change View
25 MG
$180.00
100 MG
$471.25
250 MG
$873.75

About This Item

CAS Number:
Molecular Weight:
285.77
MDL number:

Vendor SKU: F20138-25MG

$180.00


Ships in 10 days - from Aldrich Partner.

Iupac Name

(4-(chloromethyl)phenyl)(3,4-dihydroisoquinolin-2(1H)-yl)methanone

Inchi Code

1S/C17H16ClNO/c18-11-13-5-7-15(8-6-13)17(20)19-10-9-14-3-1-2-4-16(14)12-19/h1-8H,9-12H2

InChI key

SQWOSHROZZKONH-UHFFFAOYSA-N

Purity

95%

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=122707

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=F20138


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service