Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATE939883205

AstaTech, Inc.

ETHYL 2-CHLORO-3-METHYL-4H-THIENO[3,2-B]PYRROLE-5-CARBOXYLATE

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$263.75
1 G
$660.00
5 G
$1,978.75

Vendor SKU: P16670-250MG

$263.75


Availability
Ships in 10 days - from Aldrich Partner.


Select a Size

Change View
250 MG
$263.75
1 G
$660.00
5 G
$1,978.75

About This Item

CAS Number:
Molecular Weight:
243.71
MDL number:

Vendor SKU: P16670-250MG

$263.75


Availability
Ships in 10 days - from Aldrich Partner.

Iupac Name

ethyl 2-chloro-3-methyl-4H-thieno[3,2-b]pyrrole-5-carboxylate

Inchi Code

1S/C10H10ClNO2S/c1-3-14-10(13)6-4-7-8(12-6)5(2)9(11)15-7/h4,12H,3H2,1-2H3

InChI key

ZFEUTGSFUAESCS-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

Pale-yellow to Yellow-brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=102361

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=P16670


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service