Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATE638636024

AstaTech, Inc.

N-BENZYL-N-(3-BROMOPYRIDIN-2-YL)PIVALAMIDE

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$263.75
250 MG
$530.00
1 G
$1,058.75

Vendor SKU: F31409-100MG

$263.75


Ships in 10 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 MG
$263.75
250 MG
$530.00
1 G
$1,058.75

About This Item

CAS Number:
Molecular Weight:
347.25
MDL number:

Vendor SKU: F31409-100MG

$263.75


Ships in 10 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.

Iupac Name

N-benzyl-N-(3-bromopyridin-2-yl)pivalamide

Inchi Code

1S/C17H19BrN2O/c1-17(2,3)16(21)20(12-13-8-5-4-6-9-13)15-14(18)10-7-11-19-15/h4-11H,12H2,1-3H3

InChI key

DCIRVBJXKJONOC-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

Pale-yellow to Yellow-brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=97511

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=F31409


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service