Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATE514315597

AstaTech, Inc.

TERT-BUTYL (2-CHLOROPYRIMIDIN-4-YL)(METHYL)CARBAMATE

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$248.75
250 MG
$558.75

Vendor SKU: W10951-100MG

$248.75


Ships in 10 days - from Aldrich Partner.


Select a Size

Change View
100 MG
$248.75
250 MG
$558.75

About This Item

CAS Number:
Molecular Weight:
243.69
MDL number:

Vendor SKU: W10951-100MG

$248.75


Ships in 10 days - from Aldrich Partner.

Iupac Name

tert-butyl (2-chloropyrimidin-4-yl)(methyl)carbamate

Inchi Code

1S/C10H14ClN3O2/c1-10(2,3)16-9(15)14(4)7-5-6-12-8(11)13-7/h5-6H,1-4H3

InChI key

ZNTAQUJSTLQNPT-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Colorless to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=80147

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=W10951


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service