Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATE456786563

AstaTech, Inc.

2-METHOXY-5-(TRIFLUOROMETHOXY)BENZONITRILE

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$43.75
5 G
$160.00

Vendor SKU: CL9481-1G

$43.75


Availability
Ships in 10 days - from Aldrich Partner.


Select a Size

Change View
1 G
$43.75
5 G
$160.00

About This Item

CAS Number:
Molecular Weight:
217.15
MDL number:

Vendor SKU: CL9481-1G

$43.75


Availability
Ships in 10 days - from Aldrich Partner.

Iupac Name

2-methoxy-5-(trifluoromethoxy)benzonitrile

Inchi Code

1S/C9H6F3NO2/c1-14-8-3-2-7(4-6(8)5-13)15-9(10,11)12/h2-4H,1H3

InChI key

FJFWRXJVLWLALW-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Yellow to Brown Liquid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=65857

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=CL9481


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service