Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATE456785831

AstaTech, Inc.

5-BROMO-2-(TRIFLUOROMETHYL)THIAZOLE

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$250.00
250 MG
$523.75
1 G
$1,247.50

Vendor SKU: 79148-50MG

$250.00


Ships in 10 days - from Aldrich Partner.


Select a Size

Change View
50 MG
$250.00
250 MG
$523.75
1 G
$1,247.50

About This Item

CAS Number:
Molecular Weight:
232.02
MDL number:

Vendor SKU: 79148-50MG

$250.00


Ships in 10 days - from Aldrich Partner.

Iupac Name

5-bromo-2-(trifluoromethyl)-1,3-thiazole

Inchi Code

1S/C4HBrF3NS/c5-2-1-9-3(10-2)4(6,7)8/h1H

InChI key

BEFIZUOVAXMSPV-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Colorless to Yellow Liquid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=64813

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=79148


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service