Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATE448437107

AstaTech, Inc.

TERT-BUTYL 4-((TRIMETHYLSILYL)ETHYNYL) BENZOATE

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$328.75
1 G
$821.25

Vendor SKU: AB10916-250MG

$328.75


Ships in 10 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
250 MG
$328.75
1 G
$821.25

About This Item

CAS Number:
Molecular Weight:
274.43
MDL number:

Vendor SKU: AB10916-250MG

$328.75


Ships in 10 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.

Iupac Name

tert-butyl 4-[(trimethylsilyl)ethynyl]benzoate

Inchi Code

1S/C16H22O2Si/c1-16(2,3)18-15(17)14-9-7-13(8-10-14)11-12-19(4,5)6/h7-10H,1-6H3

InChI key

KAYVQNMVOJPPLD-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Yellow to Brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=59205

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=AB10916


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service