Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATE448435178

AstaTech, Inc.

2-(5-CHLORO-2-THIENYL)PYRROLIDINE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
187.69
MDL number:

Iupac Name

2-(5-chloro-2-thienyl)pyrrolidine

Inchi Code

1S/C8H10ClNS/c9-8-4-3-7(11-8)6-2-1-5-10-6/h3-4,6,10H,1-2,5H2

InChI key

XMSYAIXTGCQRQX-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

97%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Yellow to Brown Liquid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=61787

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=77444


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service