Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATE448433804

AstaTech, Inc.

4-ISOPROPOXYBENZENESULFONYL CHLORIDE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
234.7
MDL number:

bp

112-113 C/0.01 mmHg

mp

30-37 C

Iupac Name

4-isopropoxybenzenesulfonyl chloride

Inchi Code

1S/C9H11ClO3S/c1-7(2)13-8-3-5-9(6-4-8)14(10,11)12/h3-7H,1-2H3

InChI key

IJWCRHKAQNFJLT-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

97%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=60441

Boiling point

112-113 C/0.01 mmHg

Melting Point

30-37 C

pictograms

Signal Word

Danger

Hazard Statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=76143


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service