Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATE448432446

AstaTech, Inc.

ETHYL 2-(4-BENZYLPIPERAZIN-2-YL)ACETATE

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$250.00
50 MG
$748.75
250 MG
$2,496.25

Vendor SKU: 74791-10MG

$250.00


Ships in 10 days - from Aldrich Partner.


Select a Size

Change View
10 MG
$250.00
50 MG
$748.75
250 MG
$2,496.25

About This Item

CAS Number:
Molecular Weight:
262.35
MDL number:

Vendor SKU: 74791-10MG

$250.00


Ships in 10 days - from Aldrich Partner.

Iupac Name

ethyl 2-(4-benzylpiperazin-2-yl)acetate

Inchi Code

1S/C15H22N2O2/c1-2-19-15(18)10-14-12-17(9-8-16-14)11-13-6-4-3-5-7-13/h3-7,14,16H,2,8-12H2,1H3

InChI key

JISNWYDETMHNFI-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

90%

Shipping Temperature

Normal

Physical Form

Pale-yellow to Yellow-brown Liquid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=58735

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=74791


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service