Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATE443542572

AstaTech, Inc.

4-FLUOROCINNAMONITRILE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
147.15
MDL number:

bp

65-75 C/0.2 mmHg

mp

69-70 C

Iupac Name

(2E)-3-(4-fluorophenyl)-2-propenenitrile

Inchi Code

1S/C9H6FN/c10-9-5-3-8(4-6-9)2-1-7-11/h1-6H/b2-1+

InChI key

WVWTVBZOOBKCKI-OWOJBTEDSA-N

storage temp.

2-8 C

Purity

97%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=51649

Boiling point

65-75 C/0.2 mmHg

Melting Point

69-70 C

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=36939


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service