Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATE423890471

AstaTech, Inc.

METHYL 3-NITROISONICOTINATE

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$130.00
1 G
$473.75
5 G
$1,846.25

Vendor SKU: 36497-250MG

$130.00


Ships in 10 days - from Aldrich Partner.


Select a Size

Change View
250 MG
$130.00
1 G
$473.75
5 G
$1,846.25

About This Item

CAS Number:
Molecular Weight:
182.14
MDL number:

Vendor SKU: 36497-250MG

$130.00


Ships in 10 days - from Aldrich Partner.

Iupac Name

methyl 3-nitroisonicotinate

Inchi Code

1S/C7H6N2O4/c1-13-7(10)5-2-3-8-4-6(5)9(11)12/h2-4H,1H3

InChI key

VFZBLITUPLITGH-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Yellow to Brown Liquid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=36799

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=36497


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service