Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATE423889975

AstaTech, Inc.

TERT-BUTYL 4-(4-CHLOROBENZOYL)PIPERAZINE-1-CARBOXYLATE

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$120.00

Vendor SKU: 36651-1G

$120.00


Ships in 10 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
1 G
$120.00

About This Item

CAS Number:
Molecular Weight:
324.81
MDL number:

Vendor SKU: 36651-1G

$120.00


Ships in 10 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.

Iupac Name

tert-butyl 4-(4-chlorobenzoyl)-1-piperazinecarboxylate

Inchi Code

1S/C16H21ClN2O3/c1-16(2,3)22-15(21)19-10-8-18(9-11-19)14(20)12-4-6-13(17)7-5-12/h4-7H,8-11H2,1-3H3

InChI key

ZWIWAMXFONGDIO-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=36928

Signal Word

Warning

Hazard Statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=36651


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service