Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATE423888039

AstaTech, Inc.

PHENYLMETHANESULFONAMIDE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
171.22
MDL number:
Pricing and availability is not currently available. Contact Technical Service

mp

101-104 C

Iupac Name

phenylmethanesulfonamide

Inchi Code

1S/C7H9NO2S/c8-11(9,10)6-7-4-2-1-3-5-7/h1-5H,6H2,(H2,8,9,10)

InChI key

ABOYDMHGKWRPFD-UHFFFAOYSA-N

Purity

98%

Physical Form

Light-red to Brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=35333

Melting Point

101-104 C

Signal Word

Warning

Hazard Statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=33371


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service