Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATE410297852

AstaTech, Inc.

TERT-BUTYL 3-(3-AMINOPHENOXY)PIPERIDINE-1-CARBOXYLATE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
292.38
MDL number:

Iupac Name

tert-butyl 3-(3-aminophenoxy)-1-piperidinecarboxylate

Inchi Code

1S/C16H24N2O3/c1-16(2,3)21-15(19)18-9-5-8-14(11-18)20-13-7-4-6-12(17)10-13/h4,6-7,10,14H,5,8-9,11,17H2,1-3H3

InChI key

UWRYMAUCFQPCQK-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Yellow to Brown Sticky Oil to Semi-Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=22507

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=AB3309


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service