Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATE410297277

AstaTech, Inc.

TRANS-BENZYL 3-HYDROXYCYCLOPENTANECARBOXYLATE

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$533.75
1 G
$1,311.25
5 G
$3,721.25

Vendor SKU: 84941-250MG

$533.75


Ships in 10 days - from Aldrich Partner.


Select a Size

Change View
250 MG
$533.75
1 G
$1,311.25
5 G
$3,721.25

About This Item

CAS Number:
Molecular Weight:
220.27
MDL number:

Vendor SKU: 84941-250MG

$533.75


Ships in 10 days - from Aldrich Partner.

Iupac Name

benzyl (1R,3R)-3-hydroxycyclopentane-1-carboxylate

Inchi Code

1S/C13H16O3/c14-12-7-6-11(8-12)13(15)16-9-10-4-2-1-3-5-10/h1-5,11-12,14H,6-9H2/t11-,12-/m1/s1

InChI key

RDUCOWDKAANTQZ-VXGBXAGGSA-N

storage temp.

Room temperature

Purity

95%

Shipping Temperature

Normal

Physical Form

Colorless to Yellow Sticky Oil to Semi-Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=22413

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=84941


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service