Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATE398813174

AstaTech, Inc.

6-BROMOQUINAZOLINE-4-CARBOXYLIC ACID, AMMONIA SALT

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
270.09
MDL number:

Iupac Name

ammonium 6-bromo-4-quinazolinecarboxylate

Inchi Code

1S/C9H5BrN2O2.H3N/c10-5-1-2-7-6(3-5)8(9(13)14)12-4-11-7;/h1-4H,(H,13,14);1H3

InChI key

PUZDCVNKSLYUFD-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Pale-yellow to Yellow-brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=19590

Signal Word

Warning

Hazard Statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=83111


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service