Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATE371591905

AstaTech, Inc.

METHYL 3-(CBZ-AMINO)BUTANOATE

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$186.25
5 G
$641.25

Vendor SKU: 62364-1G

$186.25


Ships in 10 days - from Aldrich Partner.


Select a Size

Change View
1 G
$186.25
5 G
$641.25

About This Item

CAS Number:
Molecular Weight:
251.28
MDL number:

Vendor SKU: 62364-1G

$186.25


Ships in 10 days - from Aldrich Partner.

Iupac Name

methyl 3-{[(benzyloxy)carbonyl]amino}butanoate

Inchi Code

1S/C13H17NO4/c1-10(8-12(15)17-2)14-13(16)18-9-11-6-4-3-5-7-11/h3-7,10H,8-9H2,1-2H3,(H,14,16)

InChI key

HSKIGKMMMZQYID-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

97%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=6905

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=62364


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service