Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATE316039024

AstaTech, Inc.

3-IODO-5-METHOXY-1-METHYL-1H-INDOLE

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$250.00
250 MG
$471.25
1 G
$1,196.25

Vendor SKU: 51347-100MG

$250.00


Ships in 10 days - from Aldrich Partner.


Select a Size

Change View
100 MG
$250.00
250 MG
$471.25
1 G
$1,196.25

About This Item

CAS Number:
Molecular Weight:
287.1
MDL number:

Vendor SKU: 51347-100MG

$250.00


Ships in 10 days - from Aldrich Partner.

Iupac Name

3-iodo-5-methoxy-1-methyl-1H-indole

Inchi Code

1S/C10H10INO/c1-12-6-9(11)8-5-7(13-2)3-4-10(8)12/h3-6H,1-2H3

InChI key

UCWQLMURICCFLX-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

97%

Shipping Temperature

Normal

Physical Form

Yellow to Brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=2349

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=51347


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service