Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATE245847463

AstaTech, Inc.

2-METHYL-3-OXO-2,3-DIHYDROPYRIDAZINE-4-CARBOXYLIC ACID

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$346.25
250 MG
$821.25
1 G
$1,661.25

Vendor SKU: 63166-50MG

$346.25


Ships in 10 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
50 MG
$346.25
250 MG
$821.25
1 G
$1,661.25

About This Item

CAS Number:
Molecular Weight:
154.13
MDL number:

Vendor SKU: 63166-50MG

$346.25


Ships in 10 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.

Iupac Name

2-methyl-3-oxo-2,3-dihydro-4-pyridazinecarboxylic acid

Inchi Code

1S/C6H6N2O3/c1-8-5(9)4(6(10)11)2-3-7-8/h2-3H,1H3,(H,10,11)

InChI key

RZXOWMGNNLWTAE-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

97%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Pale-yellow to Yellow-brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=16566

Signal Word

Warning

Hazard Statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=63166


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service