Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATE067529489

AstaTech, Inc.

(2-DIISOPROPYLAMINO-ETHYL)-THIOUREA

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
203.35
MDL number:

Iupac Name

N-[2-(diisopropylamino)ethyl]thiourea

Inchi Code

1S/C9H21N3S/c1-7(2)12(8(3)4)6-5-11-9(10)13/h7-8H,5-6H2,1-4H3,(H3,10,11,13)

InChI key

XVCFOUJNSBNIJG-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

98%

Country of Origin

CN

Shipping Temperature

Normal

Product Link

http://astatechinc.com/CPSResult.php?CRNO=1292

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=37215


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service