Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APOH93ED777D

Apollo Scientific Ltd

3-Fluoro-4-(3,3,3-trifluoro-1-propyn-1-yl)benzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
231.94
MDL number:

Iupac Name

(3-fluoro-4-(3,3,3-trifluoroprop-1-yn-1-yl)phenyl)boronic acid

Inchi Code

1S/C9H5BF4O2/c11-8-5-7(10(15)16)2-1-6(8)3-4-9(12,13)14/h1-2,5,15-16H

Inchi Key

JAABBTVFYWVLOX-UHFFFAOYSA-N

Storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/3-fluoro-4-333-trifluoro-1-propyn-1-ylbenzeneboronic-acid

Pictograms

Signal Word

TBC

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/3-fluoro-4-333-trifluoro-1-propyn-1-ylbenzeneboronic-acid


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service