Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APOH93ED5E68

Apollo Scientific Ltd

Ethyl 6-bromo-2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$178.20
1 G
$534.60

Vendor SKU: PC301034-250MG

$178.20


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
250 MG
$178.20
1 G
$534.60

About This Item

CAS Number:
Molecular Weight:
319.11

Vendor SKU: PC301034-250MG

$178.20


Ships in 7 days - from Aldrich Partner.

bp

142-144

Iupac Name

ethyl 6-bromo-2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate

Inchi Code

1S/C11H9BrF2N2O2/c1-2-18-11(17)9-8(10(13)14)15-7-4-3-6(12)5-16(7)9/h3-5,10H,2H2,1H3

InChI key

WDSSODGXTHYFSK-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/ethyl-6-bromo-2-difluoromethylimidazo12-apyridine-3-carboxylate

Boiling point

142-144

pictograms

Signal Word

TBC

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/ethyl-6-bromo-2-difluoromethylimidazo12-apyridine-3-carboxylate


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service