Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APOH93DA1D1D

Apollo Scientific Ltd

Aminotriester

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
415.57
MDL number:
Pricing and availability is not currently available. Contact Technical Service

bp

60-70

Iupac Name

di(tert-butyl) 4-amino-4-(3-tert-butoxy-3-oxopropyl)heptanedioate

Inchi Code

1S/C22H41NO6/c1-19(2,3)27-16(24)10-13-22(23,14-11-17(25)28-20(4,5)6)15-12-18(26)29-21(7,8)9/h10-15,23H2,1-9H3

InChI key

PFFIXGHIRWJVRO-UHFFFAOYSA-N

storage temp.

Store at 2-8C

Purity

95%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/aminotriester

Boiling point

60-70

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/aminotriester


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service