Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APOH93D965CB

Apollo Scientific Ltd

N-Hexyltriethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
248.44
MDL number:

Iupac Name

triethoxy(hexyl)silane

Inchi Code

1S/C12H28O3Si/c1-5-9-10-11-12-16(13-6-2,14-7-3)15-8-4/h5-12H2,1-4H3

InChI key

WUMSTCDLAYQDNO-UHFFFAOYSA-N

Purity

97%

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service