Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APOH11C5AE2C

Apollo Scientific Ltd

8-Chloro-3-iodoimidazo[1,2-a]pyrazine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
279.47
MDL number:

bp

179-183

Iupac Name

8-chloro-3-iodoimidazo[1,2-a]pyrazine

Inchi Code

1S/C6H3ClIN3/c7-5-6-10-3-4(8)11(6)2-1-9-5/h1-3H

InChI key

RHQCTVNTDPLROD-UHFFFAOYSA-N

storage temp.

Store at 2-8C

Purity

97%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/8-chloro-3-iodoimidazo12-apyrazine

Boiling point

179-183

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/8-chloro-3-iodoimidazo12-apyrazine


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service