Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APOH11A9DE0C

Apollo Scientific Ltd

5-Chloro-3-cyanopyridine-2-carboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$676.50
1 G
$1,185.80

Vendor SKU: OR61368-500MG

$676.50


Ships in 7 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
500 MG
$676.50
1 G
$1,185.80

About This Item

CAS Number:
Molecular Weight:
182.57
MDL number:

Vendor SKU: OR61368-500MG

$676.50


Ships in 7 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.

bp

152

Iupac Name

5-chloro-3-cyano-2-pyridinecarboxylic acid

Inchi Code

1S/C7H3ClN2O2/c8-5-1-4(2-9)6(7(11)12)10-3-5/h1,3H,(H,11,12)

InChI key

FIQQYYHLOMZXKH-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/5-chloro-3-cyanopyridine-2-carboxylic-acid

Boiling point

152

pictograms

Exclamation mark

Signal Word

Warning

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/5-chloro-3-cyanopyridine-2-carboxylic-acid


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service