Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APOH11A9DB83

Apollo Scientific Ltd

5-(Morpholin-4-yl)-2-nitroaniline

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
223.23
MDL number:
Pricing and availability is not currently available. Contact Technical Service

Iupac Name

5-(4-morpholinyl)-2-nitroaniline

Inchi Code

1S/C10H13N3O3/c11-9-7-8(1-2-10(9)13(14)15)12-3-5-16-6-4-12/h1-2,7H,3-6,11H2

InChI key

HBBXESCZZLQWHM-UHFFFAOYSA-N

storage temp.

Store at 2-8C

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/5-morpholin-4-yl-2-nitroaniline

pictograms

Signal Word

TBC

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/5-morpholin-4-yl-2-nitroaniline


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service