Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APOH11A9CC87

Apollo Scientific Ltd

4-Bromo-3-chloro-2-fluorobenzonitrile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
234.45
MDL number:

Iupac Name

4-bromo-3-chloro-2-fluorobenzonitrile

Inchi Code

1S/C7H2BrClFN/c8-5-2-1-4(3-11)7(10)6(5)9/h1-2H

InChI key

PQFMVBRXNPGOQP-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/4-bromo-3-chloro-2-fluorobenzonitrile

pictograms

Skull and crossbones

Signal Word

Danger

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/4-bromo-3-chloro-2-fluorobenzonitrile


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service