Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APOH11A9BD95

Apollo Scientific Ltd

3,5-Bis(trifluoromethyl)-N-(bromoacetyl)aniline

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$29.70
1 G
$64.90

Vendor SKU: PC0069-250MG

$29.70


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
250 MG
$29.70
1 G
$64.90

About This Item

CAS Number:
Molecular Weight:
350.06
MDL number:

Vendor SKU: PC0069-250MG

$29.70


Ships in 7 days - from Aldrich Partner.

bp

100-101

Iupac Name

N-[3,5-bis(trifluoromethyl)phenyl]-2-bromoacetamide

Inchi Code

1S/C10H6BrF6NO/c11-4-8(19)18-7-2-5(9(12,13)14)1-6(3-7)10(15,16)17/h1-3H,4H2,(H,18,19)

InChI key

DCCKYVKUIKRMPU-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

97%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/35-bistrifluoromethyl-n-bromoacetylaniline

Synonym

3',5'-Bis(trifluoromethyl)-2-bromoacetanilide

Boiling point

100-101

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/35-bistrifluoromethyl-n-bromoacetylaniline


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service