APOH039F93C6
Apollo Scientific Ltd
Benzyl 4-cyanobicyclo[2.2.2]octan-1-ylcarbamate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
benzyl (4-cyanobicyclo[2.2.2]octan-1-yl)carbamate
Inchi Code
1S/C17H20N2O2/c18-13-16-6-9-17(10-7-16,11-8-16)19-15(20)21-12-14-4-2-1-3-5-14/h1-5H,6-12H2,(H,19,20)
InChI key
AMPXLEPLPYICLE-UHFFFAOYSA-N
storage temp.
Ambient Temperature
Country of Origin
GB
Shipping Temperature
Ambient Temperature
Physical Form
Solid
Product Link
https://store.apolloscientific.co.uk/product/benzyl-4-cyanobicyclo222octan-1-ylcarbamate
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://store.apolloscientific.co.uk/product/benzyl-4-cyanobicyclo222octan-1-ylcarbamate
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service