Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO633495407

Apollo Scientific Ltd

beta-Alanine 7-amido-4-methylcoumarin trifluoroacetate salt

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$53.90

Vendor SKU: BIB1437-50MG

$53.90


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
50 MG
$53.90

About This Item

CAS Number:
Molecular Weight:
360.29
MDL number:

Vendor SKU: BIB1437-50MG

$53.90


Ships in 7 days - from Aldrich Partner.

Iupac Name

3-amino-N-(4-methyl-2-oxo-2H-chromen-7-yl)propanamide trifluoroacetate

Inchi Code

1S/C13H14N2O3.C2HF3O2/c1-8-6-13(17)18-11-7-9(2-3-10(8)11)15-12(16)4-5-14;3-2(4,5)1(6)7/h2-3,6-7H,4-5,14H2,1H3,(H,15,16);(H,6,7)

InChI key

BFXHWJNREYBZJN-UHFFFAOYSA-N

Country of Origin

GB

Product Link

https://store.apolloscientific.co.uk/product/beta-alanine-7-amido-4-methylcoumarin-trifluoroacetate-salt

Synonym

beta-Alanine-AMC.TFA

MSDS

https://store.apolloscientific.co.uk/product/beta-alanine-7-amido-4-methylcoumarin-trifluoroacetate-salt


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service