Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO465886935

Apollo Scientific Ltd

1,1,1-Trifluoro-3-pyrrolidin-1-ylpropan-2-ol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
183.17
MDL number:

Iupac Name

1,1,1-trifluoro-3-(1-pyrrolidinyl)-2-propanol

Inchi Code

1S/C7H12F3NO/c8-7(9,10)6(12)5-11-3-1-2-4-11/h6,12H,1-5H2

InChI key

NFDYPZXRSNWUHC-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/111-trifluoro-3-pyrrolidin-1-ylpropan-2-ol

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/111-trifluoro-3-pyrrolidin-1-ylpropan-2-ol


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service