Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO456876693

Apollo Scientific Ltd

3-Fluoro-2-methylbenzoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
172.59
MDL number:
Pricing and availability is not currently available. Contact Technical Service

Iupac Name

3-fluoro-2-methylbenzoyl chloride

Inchi Code

1S/C8H6ClFO/c1-5-6(8(9)11)3-2-4-7(5)10/h2-4H,1H3

Inchi Key

OIDAXOKENZZBPF-UHFFFAOYSA-N

Storage temp.

Ambient Temperature

Purity

98%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/3-fluoro-2-methylbenzoyl-chloride

Pictograms

Signal Word

TBC

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/3-fluoro-2-methylbenzoyl-chloride


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service