Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO456794740

Apollo Scientific Ltd

5-[(1,3-Dihydro-1,3-dioxo-2H-isoindol-2-yl)methyl]-2-furaldehdye

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
255.23
MDL number:

bp

125-128

Iupac Name

5-[(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)methyl]-2-furaldehyde

Inchi Code

1S/C14H9NO4/c16-8-10-6-5-9(19-10)7-15-13(17)11-3-1-2-4-12(11)14(15)18/h1-6,8H,7H2

InChI key

ZRBXPAHJXKKXME-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/5-13-dihydro-13-dioxo-2h-isoindol-2-ylmethyl-2-furaldehdye

Boiling point

125-128

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/5-13-dihydro-13-dioxo-2h-isoindol-2-ylmethyl-2-furaldehdye


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service