Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO456794309

Apollo Scientific Ltd

1,2,3,6-Tetrahydro-2,3'-bipyridine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
160.22
MDL number:
Pricing and availability is not currently available. Contact Technical Service

density

1.086

Iupac Name

3-(1,2,3,6-tetrahydropyridin-2-yl)pyridine

Inchi Code

1S/C10H12N2/c1-2-7-12-10(5-1)9-4-3-6-11-8-9/h1-4,6,8,10,12H,5,7H2

InChI key

SOPPBXUYQGUQHE-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

94%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid-Oil

Product Link

https://store.apolloscientific.co.uk/product/1236-tetrahydro-23-bipyridine

Flash Point

116-117/1mm

density

1.086

pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/1236-tetrahydro-23-bipyridine


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service