Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO456793634

Apollo Scientific Ltd

4-Methylcyclohexen-1-ylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
139.99
MDL number:
Pricing and availability is not currently available. Contact Technical Service

bp

162-166

Iupac Name

4-methyl-1-cyclohexen-1-ylboronic acid

Inchi Code

1S/C7H13BO2/c1-6-2-4-7(5-3-6)8(9)10/h4,6,9-10H,2-3,5H2,1H3

InChI key

SJZMIZIVYIOMIW-UHFFFAOYSA-N

storage temp.

Store at -20C

Purity

98%

Country of Origin

GB

Shipping Temperature

Shipped with Ice Packs

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/4-methylcyclohexen-1-ylboronic-acid

Boiling point

162-166

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/4-methylcyclohexen-1-ylboronic-acid


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service